Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031703.1 | NP_113891.1 | 3 - 4 | 443 | 135 | |
GenBank mRNA | BC127490.1 | - | 3 - 4 | 425 | 8366493 | 135 |
D17695.1 | - | 3 - 4 | 358 | 135 | ||
L35108.1 | - | 3 - 4 | 443 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |