Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012062.1 | NP_001012062.1 | 3 - 4 | 263 | 154 | |
XM_006227617.2 | XP_006227679.1 | 2 - 3 | 296 | 154 | ||
XM_006227618.2 | XP_006227680.1 | 2 - 3 | 642 | 154 | ||
XM_006227620.2 | XP_006227682.1 | 2 - 3 | 293 | 154 | ||
XM_008758608.1 | XP_008756830.1 | 3 - 4 | 360 | 154 | ||
GenBank mRNA | BC085788.1 | - | 3 - 4 | 263 | 7108689 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |