Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012131.1 | NP_001012131.1 | 4 - 5 | 808 | 84 | |
XM_006244899.2 | XP_006244961.1 | 5 - 6 | 1028 | 84 | ||
XM_006244901.2 | XP_006244963.1 | 4 - 5 | 827 | 84 | ||
XM_008767091.1 | XP_008765313.1 | 5 - 6 | 1769 | 84 | ||
XM_008767092.1 | XP_008765314.1 | 5 - 6 | 880 | 84 | ||
XM_008767093.1 | XP_008765315.1 | 3 - 4 | 2920 | 84 | ||
GenBank mRNA | BC086600.1 | - | 4 - 5 | 808 | 7314378 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |