Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758805.1 | XP_008757027.1 | 21 - 22 | 2900 | 65 | |
XM_008758806.1 | XP_008757028.1 | 22 - 23 | 2358 | 65 | ||
XM_008758807.1 | XP_008757029.1 | 21 - 22 | 2154 | 65 | ||
XM_008758808.1 | XP_008757030.1 | 21 - 22 | 2900 | 65 | ||
XM_008758809.1 | XP_008757031.1 | 20 - 21 | 2786 | 65 | ||
XM_008758810.1 | XP_008757032.1 | 20 - 21 | 2709 | 65 | ||
XM_008758811.1 | XP_008757033.1 | 20 - 21 | 2296 | 65 | ||
XM_008758812.1 | XP_008757034.1 | 20 - 21 | 2021 | 65 | ||
XM_008758813.1 | XP_008757035.1 | 20 - 21 | 2285 | 65 | ||
XM_008758814.1 | XP_008757036.1 | 20 - 21 | 4219 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |