Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053649.1 | NP_446101.1 | 6 - 7 | 980 | 105 | |
GenBank mRNA | AB065090.1 | - | 6 - 7 | 980 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |