Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105883.1 | NP_001099353.1 | 5 - 6 | 1083 | 79 | |
XM_006248338.2 | XP_006248400.1 | 5 - 6 | 1121 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |