Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127550.1 | NP_001121022.1 | 9 - 10 | 857 | 97 | |
XM_008768735.1 | XP_008766957.1 | 7 - 8 | 761 | 97 | ||
GenBank mRNA | BC166791.1 | - | 9 - 10 | 857 | 7937077 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |