Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013161.2 | NP_037293.1 | 2 - 3 | 68 | 74 | |
GenBank mRNA | D88534.1 | - | 1 - 2 | 40 | 74 | |
D88535.1 | - | 1 - 2 | 40 | 74 | ||
M58369.1 | - | 2 - 3 | 69 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |