Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037349.2 | NP_001032426.1 | 2 - 3 | 199 | 155 | |
XM_008760400.1 | XP_008758622.1 | 2 - 3 | 226 | 155 | ||
GenBank mRNA | AY786315.1 | - | 2 - 3 | 180 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |