Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244011.2 | XP_006244073.1 | 20 - 21 | 3393 | 90 | |
XM_006244013.2 | XP_006244075.1 | 21 - 22 | 3225 | 90 | ||
XM_006244014.2 | XP_006244076.1 | 20 - 21 | 3424 | 90 | ||
XM_006244015.2 | XP_006244077.1 | 21 - 22 | 3480 | 90 | ||
XM_006244016.2 | XP_006244078.1 | 21 - 22 | 3477 | 90 | ||
XM_006244018.2 | XP_006244080.1 | 20 - 21 | 3600 | 90 | ||
XM_008766654.1 | XP_008764876.1 | 20 - 21 | 3427 | 90 | ||
XM_008766655.1 | XP_008764877.1 | 10 - 11 | 1318 | 90 | ||
XM_236654.8 | XP_236654.6 | 20 - 21 | 3634 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |