Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244188.2 | XP_006244250.1 | 1 - 2 | 173 | 97 | |
XM_008766714.1 | XP_008764936.1 | 1 - 2 | 177 | 97 | ||
GenBank mRNA | BC128697.1 | - | 1 - 2 | 119 | 7444822 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |