Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191883.1 | NP_001178812.1 | 6 - 7 | 604 | 94 | |
XM_006243802.2 | XP_006243864.1 | 7 - 8 | 1100 | 94 | ||
XM_006243803.2 | XP_006243865.1 | 7 - 8 | 1099 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |