Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276472.1 | NP_001263401.1 | 8 - 9 | 1028 | 88 | |
GenBank mRNA | FQ211656.1 | - | 8 - 9 | 1028 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |