Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168593.1 | NP_001162065.1 | 4 - 5 | 538 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |