Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012982.3 | NP_037114.2 | 1 - 2 | 382 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |