Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013030.1 | NP_037162.1 | 7 - 8 | 900 | 90 | |
GenBank mRNA | AB013453.1 | - | 7 - 8 | 900 | 90 | |
BC078876.1 | - | 7 - 8 | 894 | 7109613 | 90 | |
L13257.1 | - | 7 - 8 | 900 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |