Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191691.2 | NP_001178620.2 | 10 - 11 | 2437 | 78 | |
GenBank mRNA | AY390391.1 | - | 2 - 3 | 223 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |