Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277232.1 | NP_001264161.1 | 3 - 4 | 304 | 66 | |
NM_182953.2 | NP_891998.1 | 3 - 4 | 356 | 66 | ||
XM_006234100.1 | XP_006234162.1 | 3 - 4 | 379 | 66 | ||
GenBank mRNA | AY334013.1 | - | 2 - 3 | 253 | 66 | |
BC160820.1 | - | 2 - 3 | 380 | 7459041 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |