Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203493.3 | NP_987089.2 | 2 - 3 | 114 | 77 | |
XM_006250622.2 | XP_006250684.1 | 6 - 7 | 2687 | 77 | ||
XM_008769994.1 | XP_008768216.1 | 2 - 3 | 385 | 77 | ||
XM_008769995.1 | XP_008768217.1 | 5 - 6 | 804 | 77 | ||
GenBank mRNA | BC129082.1 | - | 2 - 3 | 114 | 8365946 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |