Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109322.1 | NP_001102792.1 | 2 - 3 | 505 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |