Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107688.1 | NP_001101158.1 | 1 - 2 | 197 | 94 | |
XM_006232642.1 | XP_006232704.1 | 1 - 2 | 150 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |