Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244902.2 | XP_006244964.1 | 4 - 5 | 1285 | 99 | |
GenBank mRNA | FQ222344.1 | - | 5 - 6 | 1261 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |