Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109282.1 | NP_001102752.1 | 5 - 6 | 753 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |