Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035237.1 | NP_001030314.1 | 1 - 2 | 142 | 70 | |
NM_133524.2 | NP_598208.2 | 1 - 2 | 142 | 70 | ||
GenBank mRNA | AJ973227.1 | - | 1 - 2 | 142 | 70 | |
BC078750.1 | - | 1 - 2 | 70 | 7123064 | 70 | |
X54549.1 | - | 1 - 2 | 142 | 70 | ||
X62323.1 | - | 1 - 2 | 43 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |