Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007672.1 | NP_001007673.1 | 2 - 3 | 256 | 157 | |
GenBank mRNA | BC078932.1 | - | 2 - 3 | 256 | 7107480 | 157 |
FQ214419.1 | - | 2 - 3 | 275 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |