Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181632.2 | NP_853663.2 | 6 - 7 | 1055 | 87 | |
GenBank mRNA | AB109021.1 | - | 5 - 6 | 950 | 87 | |
BC084685.1 | - | 6 - 7 | 1055 | 7100793 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |