Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031344.2 | NP_112634.1 | 8 - 9 | 1142 | 142 | |
XM_008760244.1 | XP_008758466.1 | 8 - 9 | 2651 | 142 | ||
GenBank mRNA | AB021980.1 | - | 8 - 9 | 1072 | 142 | |
BC081776.1 | - | 8 - 9 | 1142 | 7128630 | 142 | |
FQ210893.1 | - | 8 - 9 | 1071 | 142 | ||
HQ909027.1 | - | 8 - 9 | 976 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |