Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012904.2 | NP_037036.1 | 9 - 10 | 772 | 94 | |
XM_006231159.1 | XP_006231221.1 | 10 - 11 | 940 | 94 | ||
XM_008760295.1 | XP_008758517.1 | 9 - 10 | 859 | 94 | ||
GenBank mRNA | BC061710.1 | - | 9 - 10 | 770 | 5598112 | 94 |
FQ216134.1 | - | 9 - 10 | 787 | 94 | ||
M19967.1 | - | 8 - 9 | 739 | 94 | ||
Y00446.1 | - | 9 - 10 | 773 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |