Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006972.1 | NP_001006973.1 | 8 - 9 | 1170 | 101 | |
GenBank mRNA | BC083709.1 | - | 8 - 9 | 1170 | 7190583 | 101 |
FQ214688.1 | - | 8 - 9 | 1178 | 101 | ||
FQ217076.1 | - | 8 - 9 | 1228 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |