Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024888.1 | NP_001020059.1 | 6 - 7 | 1105 | 63 | |
XM_008761144.1 | XP_008759366.1 | 6 - 7 | 1036 | 63 | ||
GenBank mRNA | BC097428.1 | - | 6 - 7 | 1105 | 7455294 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |