Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134409.1 | NP_001127881.1 | 4 - 5 | 873 | 115 | |
XM_008761077.1 | XP_008759299.1 | 5 - 6 | 1046 | 115 | ||
GenBank mRNA | X99773.1 | - | 4 - 5 | 888 | 115 | |
Z30585.1 | - | 2 - 3 | 241 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |