Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011950.2 | NP_001011950.2 | 1 - 2 | 317 | 109 | |
XM_006232610.2 | XP_006232672.1 | 1 - 2 | 113 | 109 | ||
XM_006232611.2 | XP_006232673.1 | 1 - 2 | 113 | 109 | ||
GenBank mRNA | BC085878.1 | - | 1 - 2 | 317 | 7191747 | 109 |
FQ230674.1 | - | 1 - 2 | 107 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |