Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022202.1 | NP_071538.1 | 1 - 2 | 580 | 108 | |
GenBank mRNA | U63288.1 | - | 1 - 2 | 580 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |