Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032077.1 | NP_114466.1 | 3 - 4 | 210 | 97 | |
GenBank mRNA | BC091403.1 | - | 3 - 4 | 251 | 7324332 | 97 |
CO570909.1 | - | 3 - 4 | 210 | 97 | ||
FQ209439.1 | - | 3 - 4 | 228 | 97 | ||
FQ211027.1 | - | 3 - 4 | 221 | 97 | ||
FQ211041.1 | - | 3 - 4 | 264 | 97 | ||
FQ218360.1 | - | 3 - 4 | 225 | 97 | ||
FQ218888.1 | - | 3 - 4 | 221 | 97 | ||
FQ219478.1 | - | 3 - 4 | 213 | 97 | ||
FQ219506.1 | - | 3 - 4 | 221 | 97 | ||
FQ219654.1 | - | 3 - 4 | 213 | 97 | ||
FQ219776.1 | - | 3 - 4 | 221 | 97 | ||
FQ223824.1 | - | 2 - 3 | 121 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |