Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031730.2 | NP_113918.2 | 2 - 3 | 1831 | 84 | |
XM_008762830.1 | XP_008761052.1 | 2 - 3 | 355 | 84 | ||
GenBank mRNA | CO393575.1 | - | 2 - 3 | 392 | 84 | |
M59980.1 | - | 2 - 3 | 1510 | 84 | ||
S64320.1 | - | 2 - 3 | 1829 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |