Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134640.1 | NP_001128112.1 | 1 - 2 | 133 | 111 | |
XM_006232259.2 | XP_006232321.1 | 1 - 2 | 148 | 111 | ||
GenBank mRNA | FQ212146.1 | - | 1 - 2 | 92 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |