Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017190.4 | NP_058886.1 | 5 - 6 | 575 | 99 | |
XM_006228824.2 | XP_006228886.1 | 4 - 5 | 605 | 99 | ||
XM_006228825.2 | XP_006228887.1 | 4 - 5 | 1122 | 99 | ||
XM_006228827.2 | XP_006228889.1 | 5 - 6 | 657 | 99 | ||
XM_008759137.1 | XP_008757359.1 | 4 - 5 | 1129 | 99 | ||
XM_008759138.1 | XP_008757360.1 | 4 - 5 | 598 | 99 | ||
XM_008759139.1 | XP_008757361.1 | 4 - 5 | 1128 | 99 | ||
GenBank mRNA | FQ212027.1 | - | 5 - 6 | 591 | 99 | |
M14871.1 | - | 4 - 5 | 545 | 99 | ||
M16800.1 | - | 5 - 6 | 575 | 99 | ||
M22357.1 | - | 4 - 5 | 523 | 99 | ||
X05301.1 | - | 4 - 5 | 538 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |