Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759144.1 | XP_008757366.1 | 6 | 4030 | 89 | |
XM_008759145.1 | XP_008757367.1 | 6 | 3977 | 89 | ||
XM_008759146.1 | XP_008757368.1 | 6 | 4099 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |