Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106660.1 | NP_001100130.1 | 2 - 3 | 333 | 76 | |
GenBank mRNA | BC166422.1 | - | 2 - 3 | 253 | 9088893 | 76 |
FQ221371.1 | - | 2 - 3 | 285 | 76 | ||
FQ222881.1 | - | 2 - 3 | 247 | 76 | ||
FQ227939.1 | - | 2 - 3 | 285 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |