Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232612.2 | XP_006232674.1 | 3 - 4 | 1317 | 75 | |
XM_008761135.1 | XP_008759357.1 | 3 - 4 | 1329 | 75 | ||
GenBank mRNA | BC099090.1 | - | 3 - 4 | 602 | 75 | |
FQ216274.1 | - | 3 - 4 | 582 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |