Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013035.1 | NP_001013053.1 | 3 - 4 | 421 | 100 | |
GenBank mRNA | BC083705.1 | - | 3 - 4 | 421 | 7190294 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |