Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763338.1 | XP_008761560.1 | 3 - 4 | 327 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |