Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237697.2 | XP_006237759.1 | 6 - 7 | 1880 | 82 | |
XM_575726.6 | XP_575726.2 | 6 - 7 | 1177 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |