Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763345.1 | XP_008761567.1 | 6 - 7 | 2088 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |