Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763231.1 | XP_008761453.1 | 1 - 2 | 1396 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |