Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037189.1 | NP_001032266.1 | 2 - 3 | 120 | 139 | |
GenBank mRNA | BC099120.1 | - | 2 - 3 | 120 | 7462721 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |