Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 11 - 12 | 1693 | 58 | |
XM_006245977.2 | XP_006246039.1 | 11 - 12 | 5511 | 58 | ||
XM_008767578.1 | XP_008765800.1 | 12 - 13 | 4628 | 58 | ||
XM_008767579.1 | XP_008765801.1 | 12 - 13 | 4503 | 58 | ||
XM_008767580.1 | XP_008765802.1 | 11 - 12 | 2613 | 58 | ||
GenBank mRNA | BC099777.1 | - | 11 - 12 | 1693 | 7933232 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |