Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100158.2 | NP_001093628.1 | 4 - 5 | 436 | 69 | |
GenBank mRNA | BC105875.1 | - | 4 - 5 | 381 | 7130292 | 69 |
CO559861.1 | - | 4 - 5 | 436 | 69 | ||
FQ234161.1 | - | 3 - 4 | 157 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |