Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107666.2 | NP_001101136.2 | 6 - 7 | 908 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |