Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014113.2 | NP_001014135.2 | 6 - 7 | 570 | 98 | |
GenBank mRNA | BC099799.1 | - | 6 - 7 | 570 | 7936638 | 98 |
FQ227994.1 | - | 6 - 7 | 557 | 98 | ||
FQ231634.1 | - | 6 - 7 | 561 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |